Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ NOVAX™Electrical Insulation Gloves

Offer strength and durability

Manufacturer:  PIP™ 1470011/10

 View more versions of this product

Catalog No. 19-168-2072


Add to cart

Description

Description

Ideal for use in utilities, industrial plant maintenance, hybrid automotive maintenance/repair/service, electrical contracting, and power generation.

  • Natural rubber construction ensures the required dielectric properties combined with flexibility
  • Contoured shape reduces hand fatigue

Compliance

Gloves are individually tested to meet ANSI™/ASTM™ D120 Standard and NFPA 70E for use around electrical hazards and arc flash protection .

Specifications

Specifications

Yes
11 x 4.7 in. (27.9 x 12cm)
Rolled
Rubber, natural
10
4.7 in.
0.75mm
11 in.
ISO9001:2003
00
Smooth
exclusively for electrical purposes, such as power generation, electrical contractors, hybrid automotive repair/service, hybrid automobile manufacturing, industrial plant maintenance, utilities
Contoured
500VAC, 750VDC
12cm
Orange
27.9cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit