Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ NOVAX™ Class 0 Natural Rubber Electrical Insulating Gloves - 11 in.

Catalog No. 191682118 Shop All Protective Industrial Products Inc. Products
Click to view available options
Color:
Black
Orange
Size:
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves are constructed of natural rubber to provide dielectric protection, and compliant with ANSI/ASTM D120 and NFPA 70E standards for use around electrical hazards.

  • Class 0 electrical insulating gloves constructed of natural rubber
  • Flexible material allows workers to easily handle small, delicate parts
  • Contoured shape is comfortable and helps to reduce hand fatigue
  • Gloves meet or exceed all requirements of ANSI/ASTM D120 and NFPA 70E
  • Ideal for use around electrical and arc flash hazards
  • Individually numbered for traceability
  • Stamped with factory manufacturing date
  • Silicone-free

Specifications

Certifications/Compliance ANSI/ASTM D120, Class 0, Type I, NFPA 70E
Applications Industrial
Color Black
Exterior Finish No Texture
Hand Specific
Material Natural Rubber
Product Type Lineman's Gloves
Size 10
For Use With (Application) Electrical Utility Work, Hybrid Automobiles, Industrial Plant Maintenance, and Power Generation
Glove Cuff Style Straight
Series 150-0-11
Length (English) 11 in.
Length (Metric) 28 cm
Voltage 1000 V
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.