Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Heavy Weight Seamless Knit Cotton/Polyester Gloves with Double-Sided PVC Dot Grip

Catalog Number p-3347698
Click to view available options
Size:
Small
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Seamless knit shell is comfortable and flexible, while the PVC dots & coated fingertips enhance grip capabilities and abrasion resistance.

  • Double-sided PVC dots provide enhance grip, while also providing cut and abrasion resistance
  • Coated finger tips extend glove life, and increase durability in high-wear areas
  • Seamless knit shell is comfortable and flexible
  • Knit wrist helps prevent dirt and debris from entering the glove
Construction Seamless Knit
Coating Material PVC Dots (2 Sides)
Color Natural
Material Polycotton
Product Type Seamless Knit Gloves
Size Small
For Use With (Application) Warehousing, General Assembly, Construction, and Bottle Handling
Glove Cuff Style Continuous Knit
Length (English) 8.9 in.
Length (Metric) 22.5 cm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit