Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ PermaFlex™ Cold-Resistant PVC-Coated Gloves

Gloves stay soft and flexible in cold applications

Manufacturer:  PIP™ 588651/S

 View more versions of this product

Catalog No. 19-175-106


Add to cart

Description

Description

PVC-coated gloves are water- and cold-resistant — ideal for cold-water marine applications.

  • Seamless liner provides excellent tactile sensitivity
  • Treated to resist germs and odors
  • Rough-coated palm for better grip
  • 25.4cm (10 in.): Brown
  • 30.5cm (12 in.): Orange

Compliance

Meets EN 388 - 3121 standards.

Specifications

Specifications

11.8 x 4 in. (30 x 10.2cm)
Pinked
PVC (Polyvinyl Chloride)
Small
10.2cm
11.8 in.
Rough
application where machine Oil is present and for applications that require a combination of liquid and cut protection
Cotton
4 in.
Orange
30cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit