Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ PowerGrab Premium Gloves with Latex MicroFinish™ Grip

Superior grip and comfort

Manufacturer:  PIP™ 39C1500/L

 View more versions of this product

Catalog No. 19-165-736D


Add to cart

Description

Description

Latex MicroFinish coating provides a superior grip in dry, wet and oily conditions by reacting like tiny suction cups that attach themselves firmly to the material being handled.

  • Seamless cotton/polyester knit shell is comfortable and flexible
  • Knit wrist helps prevent dirt and debris from entering the glove
  • Ergonomic shaped curved finger pattern
  • Breathable back for comfort
  • EN Cut Level 2
  • Color: Brown/black

Specifications

Specifications

Palm, finger
Recommend washing gloves in tempered water not exceeding 40°C, a mild commercial laundry soap or detergent should be used, wash in a 5 to 10 min. cycle time, rinse in cold water, hang dry
MicroFinish™
Beige
Cotton, Polyester
4.3 in.
Brown
25.4 cm
Level 2
25.4 x 10.8 cm (10 x 4.3 in.)
Continuous knit
Rubber
Large
10.8 cm
10 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit