Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ PowerGrab Thermo Hi-Vis Orange Gloves

Excellent thermal insulation

Manufacturer:  Protective Industrial Products, Inc. 411400/L

 View more versions of this product

Catalog No. 19-166-1221



  Check Availability
Add to cart

Alert:

This style contains natural rubber latex which may cause allergic reactions. Safe use by or on latex-sensitive individuals has not been established.


Specifications Description & Specifications

Specifications

10.2 x 4.1 in. (26 x 10.5cm)
Tan
Acrylic Terry,Latex,MicroFinish™
4.1 in.
Orange
26cm
Continuous knit
refrigerated areas, utilities, food processing, commercial fishing, construction, outdoor winter activities and recreation
Large
10.5cm
10.2 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit