Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ProCoat™ Smooth PVC-Coated Hi-Vis Insulated Gloves - Safety Cuff

Click to view available options
One Size
This item is not returnable. View return policy
This item is not returnable. View return policy

Liquid-proof gloves feature foam insulation for protection against cold temperatures. Hi-vis orange color increases visibility in construction zones and warehouses.

  • Smooth, liquid-proof PVC coating covers the entire glove
  • Foam insulation helps keep hands warm and dry
  • Safety cuff helps to protect the wrist and allows for fast removal
  • Hi-vis orange color increases worker safety in warehouses and on the highway
  • Ideal for handling grease, oils, acids, caustics, and solvents
Color Hi-Vis Orange
Exterior Finish No Texture
Hand Specific
Material PVC
Size Universal
For Use With (Application) Highway Construction, Traffic Control, and Warehousing
Gender Men's
Glove Cuff Style Safety
Series ProCoat™
Length (English) 10.6 in.
Length (English) Cuff 2.8 in.
Length (Metric) 27 cm
Length (Metric) Cuff 7 cm
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov