Learn More
PIP™ ProCoat Single-Dip PVC Glove
Used for mining, construction, refining, petrochemicals and handling oily materials.
Manufacturer:  PIP™ 588060
Catalog No. 19-120-1472
Add to cart
Description
Description
  • Interlock lining dries quickly, sheds little lint and offers great dexterity
  • Black color styles hide dirt
  • Single dip construction
  • Straight cuff adds additional length to protect the forearm from liquid run-off
Specifications
Specifications
CE, EN 388: 4121
Gauntlet
Interlock knit
Men's
18 in.
Smooth
mining, construction, refining, Petrochemicals and handling Oily materials
PVC (Polyvinyl Chloride)
Black
45.7cm