Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Maximum Safety™ Leather Palm Blue Mechanics Gloves

Catalog Number p-3080845
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Designed to protect the professional mechanic or enthusiast from heat, cold, cuts, and abrasions. The washable Amara™ synthetic leather palm and thumb extend the glove's useful life.

  • Spandex back provides a snug fit for increased grip & finger control
  • Double stitched to provide additional durability
  • Hook & Loop closure for a secure fit and easy on/off
  • Launderable fabric extended life and reduces replacement costs
  • Ideal for automotive, aerospace, or general industrial maintenance
Back Material Spandex
Coating Material Uncoated
Color Blue, Black
Hand Specific
Material Synthetic Leather
Product Type Mechanic's Gloves
Size Medium
For Use With (Application) Automotive Repairs, Aerospace, or General Industrial Maintenance
Glove Cuff Style Hook and Loop
Series 120-MX2830
Length (English) 8.7 in.
Length (Metric) 22.2 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit