Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Professional Mechanic's Gloves

Designed to protect from heat, cold, cuts and abrasions

Manufacturer:  Protective Industrial Products, Inc. 120MX2830/XL

 View more versions of this product

Catalog No. 19-160-820D


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Amara™ Synthetic Leather, Cotton, Polyester
automotive, aerospace or general industrial maintenance
X-Large
11.5cm
Black,Blue
23.4cm
9.2 x 4.5 in. (23.4 x 11.5cm)
Cotton,Leather, synthetic,Polyester
4.5 in.
ISO9000:2007
9.2 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit