Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ PVC Coated Cotton/Polyester Gloves

Comfortable and flexible

Manufacturer:  Protective Industrial Products, Inc. 37C110PCBL/L

 View more versions of this product

Catalog No. 19-121-228B


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Palm
10 x 4.3 in. (25.4 x 10.8cm)
Continuous knit
warehousing and package handling, general assembly, light duty general work situations, construction and bottle handling
Cotton, Polyester
4.3 in.
Blue
25.4cm
Recommend washing gloves in slightly warm water not exceeding 40°C, a mild commercial laundry soap or detergent should be used, wash in a 5 to 10 min. cycle time, rinse in cold water, tumble dry at temperature not exceeding 60°C
Seamless
White
PVC (Polyvinyl Chloride)
Large
10.8cm
10 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit