Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Split-Cowhide Drivers

Premium Grade. Used in cold weather applications including farming, construction, machine operation and refrigeration.

Manufacturer:  PIP™ 77289TL/L

 View more versions of this product

Catalog No. 19-120-1597C

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Split Cowhide leather construction provides comfort, durability, excellent abrasion resistance, and breathability
  • Split leather is economical and has a visible texture
  • 3M™ Thinsulate™ lining provides superior insulation without the bulk of fleece
  • unique microfibers or fine fibers that make up Thinsulate insulation work by trapping air molecules between you and the outside
  • Drivers gloves feature slip-on styling to provide good fit and comfort
  • Keystone thumb is ergonomic, reduces hand fatigue, offers superior dexterity and durability
  • Elastic closure sewn inside glove for safety and comfort
  • Color coded hems for easy size identification

Order Info

Sold 12 pairs per pack.

Specifications

Specifications

10.4 x 5.5 in. (26.5 x 14cm)
cold weather applications including farming, construction, machine operation and refrigeration
Thinsulate™
5.5 in.
0.9mm
10.4 in.
Rolled
Leather, cowhide
Large
14cm
Beige
26.5cm
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit