Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Top-Grain, Quality-Grade Cowhide Driver's Gloves with Wing Thumb

Superior Grade. Used in construction, machine operation, light duty welding, utility work, farming, roping and many other general applications.

$87.00 - $90.00

Specifications

Color Natural
Material Cowhide Leather
For Use With (Application) Construction, machine operation, light duty welding, utility work, farming, roping, and many other general applications
Glove Cuff Style Slip-on
Thickness (Metric) 1 to 1.1 mm
View More Specs

Products 4
Catalog Number Mfr. No. Length (English) Size Hem Color Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Size Hem Color Width (English) Price Quantity & Availability  
19-130-0537C
View Documents
PIP™
68116/L
9.8 in. Large Brown 5 in.
Dozen of 12 for $87.00
Add to cart
 
19-130-0537A
View Documents
PIP™
68116/S
8.8 in. Small Red 4.5 in.
Dozen of 12 for $87.00
Add to cart
 
19-130-0537B
View Documents
PIP™
68116/M
9.3 in. Medium Green 4.8 in.
Dozen of 12 for $87.00
Add to cart
 
19-130-0537D
View Documents
PIP™
68116/XL
10.3 in. X-Large Blue 5.3 in.
Dozen of 12 for $90.00
Add to cart
 
Description

Description

  • Top Grain Cowhide leather construction provides comfort, durability, excellent abrasion resistance, and breathability
  • Top Grain leather is smooth, provides durability and dexterity
  • Stitched with Kevlar™, adding strength and burn resistance
  • Drivers gloves feature slip-on styling to provide good fit and comfort
  • Wing thumb increases comfort in open-handed work
  • Elastic closure sewn inside glove for safety and comfort
  • Color coded hems for easy size identification

Specifications

Specifications

Natural
Construction, machine operation, light duty welding, utility work, farming, roping, and many other general applications
1 to 1.1 mm
Cowhide Leather
Slip-on
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit