Learn More
PIP™ TOWA™ ActiveGrip Advance™ Nitrile-Coated Gloves
Revolutionary new surface treatment actively grips object being held
Manufacturer: PIP™ 34AG581/L
Description
Unlike rough surface treatments, MicroFinish™ adjusts to different conditions by reacting like tiny suction cups that attach themselves firmly to the material being handled. Used in general assembly of oily parts, metal handling, fishing and machine operation. Black.
- Provide superior grip in dry, wet, and oily conditions
- Exhibit excellent resistance to liquid permeation
- Offer strong protection against punctures and abrasions
- Seamless nylon liner promotes comfort and better fit
- Liner is treated to resist germs and odors
- Open back provides comfort
Specifications
Large |