Learn More
PIP™ Xtra Tuff™ PVC Oil-Resistant Gloves
Great for applications where machine oil is present and for applications that require a combination of liquid and cut protection.
Manufacturer:  PIP™ 588658K/L
Catalog No. 19-120-354
Add to cart
Description
Description
  • Dupont™ Kevlar™ fiber construction provides high tensile strength relative to its weight, is inherently cut resistant and will not melt, ignite or conduct electricity
  • Oil and liquid-resistant, staying soft and strong even after exposure to machine oils
  • Seamless knit construction protects hands without sacrificing comfort or dexterity
  • Straight cuff adds additional length to protect the forearm from liquid run-off
  • Rough coated palm provides a great grip
  • Seamless Kevlar liner provides cut resistance, excellent tactile sensitivity and is treated to resist germs and odors
  • Kevlar is light, comfortable and provides enduring protection against heat, cuts, lacerations and abrasions
Specifications
Specifications
Seamless
Kevlar™, PVC Coated
Open
Kevlar™, PVC (Polyvinyl Chloride)
4.9 in.
Blue
30.5cm
Level 4
12 x 4.9 in. (30.5 x 12.5cm)
where machine Oil is present and for applications that require a combination of liquid and cut protection
Large
12.5cm
12 in.