Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP4K2A Mouse anti-Human, Clone: OTI1B2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525387

Catalog No. PIMA525387


Add to cart

Description

Description

Phosphatidylinositol-5,4-bisphosphate, the precursor to second messengers of the phosphoinositide signal transduction pathways, is thought to be involved in the regulation of secretion, cell proliferation, differentiation, and motility. The protein encoded by this gene is one of a family of enzymes capable of catalyzing the phosphorylation of phosphatidylinositol-5-phosphate on the fourth hydroxyl of the myo-inositol ring to form phosphatidylinositol-5,4-bisphosphate. The amino acid sequence of this enzyme does not show homology to other kinases, but the recombit protein does exhibit kinase activity. This gene is a member of the phosphatidylinositol-5-phosphate 4-kinase family.
Specifications

Specifications

PIP4K2A
Monoclonal
0.46 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P48426
Phosphatidylinositol 5-phosphate 4-kinase type-2 alpha; 1-phosphatidylinositol 5-phosphate 4-kinase 2-alpha; Diphosphoinositide kinase 2-alpha; PIP5KIII; Phosphatidylinositol 5-phosphate 4-kinase type II alpha; PI(5)P 4-kinase type II alpha; PIP4KII-alpha; PtdIns(4)P-5-kinase B isoform; PtdIns(4)P-5-kinase C isoform; PtdIns(5)P-4-kinase isoform 2-alpha; PIP4K2A; PIP5K2; PIP5K2A
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5305
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1B2
Unconjugated
PIP4K2A
Liquid
PIP4K2A
Full length human recombit protein of PIP4K2A produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit