Learn More
Abnova™ PIP5K1B (Human) Recombinant Protein
For use with Functional Study.
Manufacturer:  Abnova™ P6575
Catalog No. 89-339-437
Add to cart
Description
Description
Human PIP5K1B (NP_003549.1, 1 a.a. - 540 a.a.) full length recombinant protein with GST-tag at N-terminal using baculovirus expression system.
Specifications
Specifications
Functional Study
phosphatidylinositol-4-phosphate 5-kinase, type I, beta
5 µg
MSS4, STM7
8395
The purity was assessed by SDS-PAGE/CBB staining.
Stored at -80°C. Aliquot to avoid repeated freezing and thawing.