Learn More
Cole-Parmer Masterflex Fitting, PVDF, Straight, Hose Barb to Threaded Adapter, 3/4" ID x 3/4" NPT(M); 5/PK  DFS Item
Manufacturer:  Cole-Parmer 3070424
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi). PVDF material features excellent chemical resistance with a wide temperature range. Sterilize via ethylene oxide or autoclave.
Catalog No. 50-628-729