Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Pipe Marker for Gas, Legend: FULL CYLINDERS READY FOR USE

For identification of pipe content. Brady chemical and hazardous material signs pipe markers for gases are made from pressure sensitive vinyl with permanent adhesive.

Manufacturer:  Brady™ 17953

Catalog No. 19-112-061


Add to cart

Specifications

Specifications

Acrylic, Permanent, Cold temperature
Legend: FULL CYLINDERS READY FOR USE
W x H: 10 x 7 in.
0.004 in.
Pipe marker for gases
White, Green
Abrasion and chemical resistant
W x H: 25.4 x 17.7cm
Pressure Sensitive Vinyl (B-946)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit