Learn More
Cole-Parmer Masterflex Fitting, HDPE, Straight, Threaded Plug, 3/4" NPT(M); 100/PK  DFS Item
Manufacturer:  Cole-Parmer 3040149
Very good chemical resistance. Rated to 200 psi at 77ºF ( 25ºC). Usable from -65 to 190ºF (-53 to 87ºC).
Catalog No. 50-630-169