Learn More
Cole-Parmer Masterflex Fitting, Nylon, Straight, Threaded Plug, 3/8" NPT(M); 10/PK  DFS Item
Manufacturer:  Cole-Parmer 3040150
Very good chemical resistance. Rated to 200 psi at 77ºF ( 25ºC). Usable from -65 to 190ºF (-53 to 87ºC).
Catalog No. 50-630-185