Learn More
Cole Parmer Cole-Parmer Pipe Fittings, Pipe plugs NPT(M), Kynar, 1/8", 5/8", 17/32"; 10/Pk DFS Item
Manufacturer:  Cole Parmer 3070621
Offering very good chemical resistance, these plugs are rated to 200 psi at 77ºF ( 25ºC). Usable from -65 to 190ºF (-53 to 87°C).
Catalog No. NC0151875