Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Piperacillin, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mg
1 g
Description
A semisynthetic penicillin with wide spectrum of antimicrobial activity, particularly pseudomonas strains.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A semisynthetic penicillin with wide spectrum of antimicrobial activity, particularly pseudomonas strains.
Solubility
Freely soluble in methanol. Only sparingly soluble in aqueous solution at 0.119 mg/mL
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
A semisynthetic penicillin with wide spectrum of antimicrobial activity, particularly pseudomonas strains.
Solubility
Freely soluble in methanol. Only sparingly soluble in aqueous solution at 0.119 mg/mL
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications
Type | Piperacillin |
Physical Form | Powder |
Melting Point | 139°C to 140°C |
Solubility Information | Freely soluble in methanol; Only sparingly soluble in aqueous solution at 0;119mg/mL |
Formula Weight | 517.55 |
Percent Purity | 95% |
Color | White |
Quantity | 500 mg |
Chemical Name or Material | Piperacillin |
Safety and Handling
Hazard Category | H317-H334 |
Hazard Statement | GHS H Statement H334-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. |
Precautionary Statement | P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c |
EINECSNumber | 262-811-8 |
RTECSNumber | XH8952200 |
TSCA | No |
storageNote1 | 2° to 8°C |
Recommended Storage | Keep cold |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction