Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Piperacillin, 95%

Click to view available options
Quantity:
500mg
1g
61477-96-1
C23H27N5O7S
517.557
MFCD00865043
IVBHGBMCVLDMKU-GXNBUGAJSA-N
43672
CHEBI:8232
(2S,5R,6R)-6-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
CCN1CCN(C(=O)C1=O)C(=O)NC(C2=CC=CC=C2)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O
61477-96-1
C23H27N5O7S
517.557
MFCD00865043
IVBHGBMCVLDMKU-GXNBUGAJSA-N
43672
CHEBI:8232
(2S,5R,6R)-6-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
CCN1CCN(C(=O)C1=O)C(=O)NC(C2=CC=CC=C2)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O
Applications
A semisynthetic penicillin with wide spectrum of antimicrobial activity, particularly pseudomonas strains.

Solubility
Freely soluble in methanol. Only sparingly soluble in aqueous solution at 0.119 mg/mL

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Solubility Information Freely soluble in methanol; Only sparingly soluble in aqueous solution at 0.119mg/mL
Formula Weight 517.55
Physical Form Powder
Percent Purity 95%
Color White
Melting Point 139°C to 140°C
Quantity 1g
Chemical Name or Material Piperacillin

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XH8952200

storageNote1 : 2° to 8°C