Learn More
Piperacillin sodium salt, 835-1007^mg/mg, Thermo Scientific Chemicals
Description
A broad spectrum antibiotic targeting a wide range of susceptible gram positive and gram negative bacteria. Piperacillin sodium salt is an antibiotic. It is used in pharmacokinetic studies to optimize antimicrobial therapy and in hypersensitivity reactions. It is involved in the study of multidrug-resistant organisms. It acts as an antibiotic related to penicillin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Soluble in ethanol, water and methanol.
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use. Store in cool place. Incompatible with acids, heavy metal salts and oxidizing agents.
Specifications
Type | Piperacillin Sodium Salt |
Physical Form | Powder |
Melting Point | 183°C to 185°C |
Beilstein | 5373920 |
Packaging | Glass Bottle |
Solubility Information | Soluble in ethanol,water and methanol. |
Formula Weight | 539.54 |
Quantity | 5 g |
Boiling Point | 793°C |
Chemical Name or Material | Piperacillin sodium salt |
Safety and Handling
Hazard Category | H317-H334 |
Precautionary Statement | P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c |
EINECSNumber | 261-868-6 |
RTECSNumber | XI0180000 |
TSCA | No |
Recommended Storage | Keep cold |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.