Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperacillin sodium salt, 835 to 1007μg/mg, Alfa Aesar™

Click to view available options
Quantity:
1g
59703-84-3
C23H26N5NaO7S
539.539
MFCD00917471
WCMIIGXFCMNQDS-IDYPWDAWSA-M
23666879
CHEBI:8233
sodium;(2S,5R,6R)-6-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CCN1CCN(C(=O)C1=O)C(=O)NC(C2=CC=CC=C2)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)[O-].[Na+]
59703-84-3
C23H26N5NaO7S
539.539
MFCD00917471
WCMIIGXFCMNQDS-IDYPWDAWSA-M
23666879
CHEBI:8233
sodium;(2S,5R,6R)-6-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CCN1CCN(C(=O)C1=O)C(=O)NC(C2=CC=CC=C2)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)[O-].[Na+]

A broad spectrum antibiotic targeting a wide range of susceptible gram positive and gram negative bacteria. Piperacillin sodium salt is an antibiotic. It is used in pharmacokinetic studies to optimize antimicrobial therapy and in hypersensitivity reactions.

Solubility
Soluble in ethanol, water and methanol.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use. Store in cool place. Incompatible with acids, heavy metal salts and oxidizing agents.
Beilstein 5373920
Odor Odorless
Merck Index 14,7463
Solubility Information Soluble in water or methanol
Formula Weight 539.54
Physical Form Powder
Boiling Point 793°C
Color White
Melting Point 183°C to 185°C
Concentration 835 to 1007μg/mg
Quantity 1g
Chemical Name or Material Piperacillin sodium salt
Show More Show Less

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XI0180000

storageNote1 : 2° to 8°C