Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperazine, 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
10kg
110-85-0
C4H10N2
86.14
MFCD00005953
GLUUGHFHXGJENI-UHFFFAOYSA-N
diethylenediamine,piperazin,1,4-diazacyclohexane,hexahydropyrazine,piperazidine,antiren,1,4-piperazine,diethyleneimine,eraverm,pipersol
4837
CHEBI:28568
piperazine
C1CNCCN1
110-85-0
C4H10N2
86.14
MFCD00005953
GLUUGHFHXGJENI-UHFFFAOYSA-N
diethylenediamine,piperazin,1,4-diazacyclohexane,hexahydropyrazine,piperazidine,antiren,1,4-piperazine,diethyleneimine,eraverm,pipersol
4837
CHEBI:28568
piperazine
C1CNCCN1
Color White to Yellow
Infrared Spectrum Authentic
Melting Point 107.0°C to 111.0°C
Quantity 100g
Water 1% max.
Flash Point 65°C
Packaging Plastic bottle
Boiling Point 146.0°C to 148.0°C
Assay Percent Range 99%
Beilstein 23,4
Fieser 04,392
Merck Index 15,7576
Solubility Information Solubility in water: 150g/L (20°C). Other solubilities: freely soluble in glycerol and glycols, soluble in alcohol, practically insoluble in diethyl ether
Formula Weight 86.14
Physical Form Crystalline Flakes
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Piperazine, anhydrous, 99%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility. Suspected of damaging the unborn child.
Flammable solid.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 203-808-3

RTECSNumber : TK7800000

TSCA : TSCA