Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperazine hexahydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
142-63-2
C4H10N2·6H2O
194.23
MFCD00149389
AVRVZRUEXIEGMP-UHFFFAOYSA-N
piperazine hexahydrate,arthriticine,piperazine, hexahydrate,vermisol,parid,unii-p3m07b8u64,usaf a-3803,diethylenediamine hexahydrate,vermyl tn
120181
piperazine;hexahydrate
C1CNCCN1.O.O.O.O.O.O
142-63-2
C4H10N2·6H2O
194.23
MFCD00149389
AVRVZRUEXIEGMP-UHFFFAOYSA-N
piperazine hexahydrate,arthriticine,piperazine, hexahydrate,vermisol,parid,unii-p3m07b8u64,usaf a-3803,diethylenediamine hexahydrate,vermyl tn
120181
piperazine;hexahydrate
C1CNCCN1.O.O.O.O.O.O
Packaging Plastic bottle
Infrared Spectrum Authentic
Flash Point 87°C
Quantity 100g
Boiling Point 145.0°C to 156.0°C
Color White
Melting Point 41.0°C to 45.0°C
Assay Percent Range 98%
Beilstein 23,4
Merck Index 14,7464
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol,practically insoluble in ether
Formula Weight 194.23
Physical Form Adhering Crystals
Percent Purity 98%
Chemical Name or Material Piperazine hexahydrate, 98%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility.
Suspected of damaging

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER

GHS Signal Word: Danger

RTECSNumber : TM0850000