Alfa Aesar™ Piperazine hydrate, ≥97%

Manufacturer: Alfa Aesar™ H370690I

Quantity 5000g

 View more versions of this product

Catalog No. AAH370690I

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 142-63-2
    Formula Weight 86.14 (Anhydrous)
    MDL Number MFCD00149389
    UN Number UN2579
    Chemical Name or Material Piperazine hydrate

    Specifications

    Physical Form Crystalline Powder or Flakes
    Formula Weight 86.14 (Anhydrous)
    Boiling Point 125° to 130°C
    Melting Point 42° to 46°C
    Health Hazard 2 GHS H Statement
    H334-H361-H314-H318-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Suspected of damaging fertility or the unborn child.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P260-P280-P303+P361+P353-P305+P351+P338-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    pH 11 to 12
    TSCA TSCA
    MDL Number MFCD00149389
    CAS 142-63-2
    UN Number UN2579
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: PIPERAZINE
    RTECS Number TM0850000
    Quantity 5000g
    Odor Weak, characteristic
    Color White
    Chemical Name or Material Piperazine hydrate
    Assay ≥97%

    GHS H Statement
    H334-H361-H314-H318-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Suspected of damaging fertility or the unborn child.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P280-P303+P361+P353-P305+P351+P338-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit