Alfa Aesar™ Piperazine hydrogen phosphate monohydrate, 98+%

Manufacturer: Alfa Aesar™ A1148736

Quantity 500g

 View more versions of this product

Catalog No. AAA1148736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18534-18-4
    Molecular Formula C4H10N2·H3PO4·H2O
    Formula Weight 202.14 (184.12 Anhydrous)
    MDL Number MFCD00156159
    Chemical Name or Material Piperazine hydrogen phosphate monohydrate

    Specifications

    Formula Weight 202.14 (184.12 Anhydrous)
    Molecular Formula C4H10N2·H3PO4·H2O
    MDL Number MFCD00156159
    CAS 18534-18-4
    Quantity 500g
    Chemical Name or Material Piperazine hydrogen phosphate monohydrate
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit