Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperine, 98%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
94-62-2
C17H19NO3
285.34
MFCD00005839
MXXWOMGUGJBKIW-YPCIICBESA-N
piperine,1-piperoylpiperidine,piperin,bioperine,piperoylpiperidine,e,e-1-piperoylpiperidine,n-e,e-piperoyl piperidine,piperidine, 1-piperoyl-, e,e,unii-u71xl721qk,fema no. 2909
638024
CHEBI:28821
(2E,4E)-5-(1,3-benzodioxol-5-yl)-1-piperidin-1-ylpenta-2,4-dien-1-one
C1CCN(CC1)C(=O)C=CC=CC2=CC3=C(C=C2)OCO3
94-62-2
C17H19NO3
285.34
MFCD00005839
MXXWOMGUGJBKIW-YPCIICBESA-N
piperine,1-piperoylpiperidine,piperin,bioperine,piperoylpiperidine,e,e-1-piperoylpiperidine,n-e,e-piperoyl piperidine,piperidine, 1-piperoyl-, e,e,unii-u71xl721qk,fema no. 2909
638024
CHEBI:28821
(2E,4E)-5-(1,3-benzodioxol-5-yl)-1-piperidin-1-ylpenta-2,4-dien-1-one
C1CCN(CC1)C(=O)C=CC=CC2=CC3=C(C=C2)OCO3
Quantity 25g
Color Tan to White or Tan-Yellow
Infrared Spectrum Authentic
Melting Point 128.0°C to 132.0°C
Packaging Glass bottle
Assay Percent Range 98%
Merck Index 15, 7584
Solubility Information Solubility in water: 40 mg/l (18°C). Other solubilities: 1g/15mL alcohol - 1g/1.7mL chloroform, 1g/36mL ether, soluble in benzene and acetic acid, practically insoluble in petroleum ether
Formula Weight 285.34
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Piperine
Show More Show Less

GHS P Statement
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Warning