Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperlongumine, Tocris Bioscience™

Catalog Number 439650
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
20069-09-4
C17H19NO5
317.34
MFCD00075706
VABYUUZNAVQNPG-BQYQJAHWSA-N
piperlongumine, piplartine, piplartin, unii-sgd66v4svj, sgd66v4svj, 5,6-dihydro-1-2e-1-oxo-3-3,4,5-trimethoxyphenyl-2-propen-1-yl-2 1h-pyridinone, e-1-3-3,4,5-trimethoxyphenyl acryloyl-5,6-dihydropyridin-2 1h-one, 1-e-3-3,4,5-trimethoxyphenyl prop-2-enoyl-2,3-dihydropyridin-6-one, e-5,6-dihydro-1-1-oxo-3-3,4,5-trimethoxyphenyl-2-propenyl-2 1h-pyridinone, 1-2e-3-3,4,5-trimethoxyphenyl prop-2-enoyl-1,2,5,6-tetrahydropyridin-2-one
637858
CHEBI:8241
1-[(2E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoyl]-1,2,5,6-tetrahydropyridin-2-one
COC1=CC(\C=C\C(=O)N2CCC=CC2=O)=CC(OC)=C1OC
This item is not returnable. View return policy
20069-09-4
C17H19NO5
317.34
MFCD00075706
VABYUUZNAVQNPG-BQYQJAHWSA-N
piperlongumine, piplartine, piplartin, unii-sgd66v4svj, sgd66v4svj, 5,6-dihydro-1-2e-1-oxo-3-3,4,5-trimethoxyphenyl-2-propen-1-yl-2 1h-pyridinone, e-1-3-3,4,5-trimethoxyphenyl acryloyl-5,6-dihydropyridin-2 1h-one, 1-e-3-3,4,5-trimethoxyphenyl prop-2-enoyl-2,3-dihydropyridin-6-one, e-5,6-dihydro-1-1-oxo-3-3,4,5-trimethoxyphenyl-2-propenyl-2 1h-pyridinone, 1-2e-3-3,4,5-trimethoxyphenyl prop-2-enoyl-1,2,5,6-tetrahydropyridin-2-one
637858
CHEBI:8241
1-[(2E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoyl]-1,2,5,6-tetrahydropyridin-2-one
COC1=CC(\C=C\C(=O)N2CCC=CC2=O)=CC(OC)=C1OC

Induces apoptosis; increases p53 and ROS levels in cancer cells

Induces cell death and increases the level of reactive oxygen species (ROS) in cancer cells with both wild-type and normal p53. Also inhibits the growth of spontaneous malignant breast tumors in mice. Displays little effect on normal cells. Rapidly depletes androgen receptor expression in human prostate cancer cells via a ROS-dependent, proteasome-mediated mechanism.
Quantity 50 mg
Formula Weight 317.34
Percent Purity >97%
Chemical Name or Material Piperlongumine
Recommended Storage Store at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit