Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperonyl alcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
495-76-1
C8H8O3
152.149
MFCD00005836
BHUIUXNAPJIDOG-UHFFFAOYSA-N
piperonyl alcohol,piperonol,1,3-benzodioxole-5-methanol,3,4-methylenedioxybenzyl alcohol,benzo d 1,3 dioxol-5-ylmethanol,piperonylalcohol,5-hydroxymethyl-1,3-benzodioxole,1-hydroxymethyl-3,4-methylenedioxybenzene,benzo 1,3 dioxol-5-ylmethanol,3,4-methylenedioxy phenylmethanol
10322
1,3-benzodioxol-5-ylmethanol
C1OC2=C(O1)C=C(C=C2)CO
495-76-1
C8H8O3
152.149
MFCD00005836
BHUIUXNAPJIDOG-UHFFFAOYSA-N
piperonyl alcohol,piperonol,1,3-benzodioxole-5-methanol,3,4-methylenedioxybenzyl alcohol,benzo d 1,3 dioxol-5-ylmethanol,piperonylalcohol,5-hydroxymethyl-1,3-benzodioxole,1-hydroxymethyl-3,4-methylenedioxybenzene,benzo 1,3 dioxol-5-ylmethanol,3,4-methylenedioxy phenylmethanol
10322
1,3-benzodioxol-5-ylmethanol
C1OC2=C(O1)C=C(C=C2)CO
Infrared Spectrum Authentic
Color White
Quantity 5g
Melting Point 51°C to 55°C
Packaging Glass bottle
Assay Percent Range 98%
Beilstein 19, 67
Formula Weight 152.15
Physical Form Crystals or Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Piperonyl alcohol
Show More Show Less