Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperonyl alcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
495-76-1
C8H8O3
152.15
MFCD00005836
BHUIUXNAPJIDOG-UHFFFAOYSA-N
piperonyl alcohol,piperonol,1,3-benzodioxole-5-methanol,3,4-methylenedioxybenzyl alcohol,benzo d 1,3 dioxol-5-ylmethanol,piperonylalcohol,5-hydroxymethyl-1,3-benzodioxole,1-hydroxymethyl-3,4-methylenedioxybenzene,benzo 1,3 dioxol-5-ylmethanol,3,4-methylenedioxy phenylmethanol
10322
1,3-benzodioxol-5-ylmethanol
C1OC2=C(O1)C=C(C=C2)CO
495-76-1
C8H8O3
152.15
MFCD00005836
BHUIUXNAPJIDOG-UHFFFAOYSA-N
piperonyl alcohol,piperonol,1,3-benzodioxole-5-methanol,3,4-methylenedioxybenzyl alcohol,benzo d 1,3 dioxol-5-ylmethanol,piperonylalcohol,5-hydroxymethyl-1,3-benzodioxole,1-hydroxymethyl-3,4-methylenedioxybenzene,benzo 1,3 dioxol-5-ylmethanol,3,4-methylenedioxy phenylmethanol
10322
1,3-benzodioxol-5-ylmethanol
C1OC2=C(O1)C=C(C=C2)CO
Color White
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 51.0°C to 55.0°C
Quantity 100g
Flash Point >110°C
Boiling Point 156.0°C to 158.0°C (66.0mmHg)
Assay Percent Range 98%
Beilstein 19, 67
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, diethyl ether, benzene,, soluble in chloroform
Formula Weight 152.15
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material Piperonyl alcohol
Show More Show Less