Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperonyl butoxide, 90%, Tech., ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass bottle
51-03-6
C19H30O5
338.44
MFCD00005842
FIPWRIJSWJWJAI-UHFFFAOYSA-N
piperonyl butoxide,butacide,butocide,ethanol butoxide,pyrenone 606,piperonylbutoxide,6-propylpiperonyl butyl diethylene glycol ether,butyl carbitol 6-propylpiperonyl ether,5-2-2-butoxyethoxy ethoxy methyl-6-propylbenzo d 1,3 dioxole,butoxide synergist
5794
CHEBI:32687
5-[2-(2-butoxyethoxy)ethoxymethyl]-6-propyl-1,3-benzodioxole
CCCCOCCOCCOCC1=CC2=C(C=C1CCC)OCO2
51-03-6
C19H30O5
338.44
MFCD00005842
FIPWRIJSWJWJAI-UHFFFAOYSA-N
piperonyl butoxide,butacide,butocide,ethanol butoxide,pyrenone 606,piperonylbutoxide,6-propylpiperonyl butyl diethylene glycol ether,butyl carbitol 6-propylpiperonyl ether,5-2-2-butoxyethoxy ethoxy methyl-6-propylbenzo d 1,3 dioxole,butoxide synergist
5794
CHEBI:32687
5-[2-(2-butoxyethoxy)ethoxymethyl]-6-propyl-1,3-benzodioxole
CCCCOCCOCCOCC1=CC2=C(C=C1CCC)OCO2
Boiling Point 180°C (1.0mmHg)
Flash Point 170°C
Viscosity 40 mPa.s (25°C)
Density 1.0600g/mL
Refractive Index 1.4970 to 1.507
Specific Gravity 1.06
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100mL
Color Yellow
Assay Percent Range 88% min. (GC)
Beilstein 19, IV, 779
Merck Index 15, 7589
Solubility Information Solubility in water: practically insoluble. Other solubilities: miscible with methanol,ethanol,benzene,freons,,and other organic solvents
Formula Weight 338.44
Physical Form Liquid
Percent Purity 90%
Grade Technical
Chemical Name or Material Piperonyl butoxide, tech., 90%
Show More Show Less

GHS H Statement
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

EINECSNumber : 200-076-7

RTECSNumber : XS8050000

TSCA : TSCA