Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperonylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
2620-50-0
C8H9NO2
151.16
MFCD00005840
ZILSBZLQGRBMOR-UHFFFAOYSA-N
piperonylamine,benzo d 1,3 dioxol-5-ylmethanamine,3,4-methylenedioxybenzylamine,1,3-benzodioxol-5-ylmethylamine,1,3-benzodioxole-5-methanamine,2h-1,3-benzodioxol-5-ylmethanamine,3,4-methylenedioxy benzylamine,benzo-1,3-dioxole-5-methylamine,1-1,3-benzodioxol-5-yl methanamine,5-aminomethyl-1,3-benzodioxole
75799
1,3-benzodioxol-5-ylmethanamine
C1OC2=C(O1)C=C(C=C2)CN
2620-50-0
C8H9NO2
151.16
MFCD00005840
ZILSBZLQGRBMOR-UHFFFAOYSA-N
piperonylamine,benzo d 1,3 dioxol-5-ylmethanamine,3,4-methylenedioxybenzylamine,1,3-benzodioxol-5-ylmethylamine,1,3-benzodioxole-5-methanamine,2h-1,3-benzodioxol-5-ylmethanamine,3,4-methylenedioxy benzylamine,benzo-1,3-dioxole-5-methylamine,1-1,3-benzodioxol-5-yl methanamine,5-aminomethyl-1,3-benzodioxole
75799
1,3-benzodioxol-5-ylmethanamine
C1OC2=C(O1)C=C(C=C2)CN
Specific Gravity 1.21
Infrared Spectrum Authentic
Color Yellow
Boiling Point 138.0°C to 139.0°C (13.0mmHg)
Density 1.2100g/mL
Flash Point >112°C
Refractive Index 1.5625 to 1.5645
Packaging Glass bottle
Quantity 25mL
Assay Percent Range 97%
Beilstein 19,I,764
Solubility Information Solubility in water: 0.9%. Other solubilities: soluble in alcohol, acetone and toluene
Formula Weight 151.16
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Piperonylamine, 97%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 220-056-1