Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperonylonitrile 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
4421-09-4
C8H5NO2
147.13
MFCD00005820
PKRWWZCDLJSJIF-UHFFFAOYSA-N
piperonylonitrile,benzo d 1,3 dioxole-5-carbonitrile,3,4-methylenedioxy benzonitrile,3,4-methylenedioxybenzonitrile,2h-1,3-benzodioxole-5-carbonitrile,benzo-1,3-dioxole-5-carbonitrile,benzonitrile, 3,4-methylenedioxy,2h-benzo d 1,3-dioxolane-5-carbonitrile,pubchem15358
78136
1,3-benzodioxole-5-carbonitrile
C1OC2=C(O1)C=C(C=C2)C#N
4421-09-4
C8H5NO2
147.13
MFCD00005820
PKRWWZCDLJSJIF-UHFFFAOYSA-N
piperonylonitrile,benzo d 1,3 dioxole-5-carbonitrile,3,4-methylenedioxy benzonitrile,3,4-methylenedioxybenzonitrile,2h-1,3-benzodioxole-5-carbonitrile,benzo-1,3-dioxole-5-carbonitrile,benzonitrile, 3,4-methylenedioxy,2h-benzo d 1,3-dioxolane-5-carbonitrile,pubchem15358
78136
1,3-benzodioxole-5-carbonitrile
C1OC2=C(O1)C=C(C=C2)C#N
Color White to Beige
Packaging Glass bottle
Melting Point 90.0°C to 95.0°C
Infrared Spectrum Authentic
Quantity 5g
Assay Percent Range 97%
Beilstein 19, 270
Formula Weight 147.13
Physical Form Crystalline Powder or Needles
Percent Purity 97%
Chemical Name or Material Piperonylonitrile, 97%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-590-6

RTECSNumber : TO2645000