Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ PIPES, 0.2M buffer soln., pH 6.8, low endotoxin

Click to view available options
Quantity:
100mL
250mL
5625-37-6
C8H18N2O6S2
302.36
MFCD00006159
IHPYMWDTONKSCO-UHFFFAOYSA-N
79723
CHEBI:44933
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
5625-37-6
C8H18N2O6S2
302.36
MFCD00006159
IHPYMWDTONKSCO-UHFFFAOYSA-N
79723
CHEBI:44933
2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O
Concentration 0.2M buffer soln.
Physical Form Liquid
pH 6.8
Color Colorless
Quantity 250mL
Chemical Name or Material PIPES, low endotoxin

TSCA : TSCA

storageNote1 : 2° to 8°C