Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIPES Disodium Salt Biological Buffer, Spectrum™ Chemical
SDP

Specifications

CAS 76836-02-7
Appearance Crystalline Powder
Physical Form Solid
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
18-621-368
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
P335225GM
Each for $78.14
N/A
18-621-369
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
P3352100GM
Each for $145.41
N/A
18-621-370
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
P33521KG
Each for $889.06
N/A
18-621-371
SDP
View Documents
Spectrum Chemical Manufacturing Corporation
P335250KGBL
50 kg
Each for $17,698.49
N/A
Description

Description

Specifications

Specifications

76836-02-7
Crystalline Powder
Solid
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit