Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIPES, Monosodium Salt, Biological Buffer, High Purity, 98.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Poly Bottle
Poly Pail
10010-67-0
10010-67-0
Packaging Poly Bottle
Loss on Drying 0.05
Quantity 25 g
Heavy Metals 15 ppm
CAS Max % 1
Percent Purity 0.985
Grade High Purity

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.