Alfa Aesar™ PIPES monosodium salt hydrate, 99%

Manufacturer: Alfa Aesar™ B21835-22

Quantity 100g

 View more versions of this product

Catalog No. AAB2183522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10010-67-0
    Molecular Formula C8H17N2NaO6S2·xH2O
    Formula Weight 324.34g/mol (Anhydrous)
    MDL Number MFCD00065472
    Merck Index 14,7479
    Synonym Piperazine-1,4-bis(ethanesulfonic acid) monosodium salt
    Chemical Name or Material PIPES monosodium salt hydrate

    Specifications

    Melting Point >300°C
    Quantity 100g
    CAS 10010-67-0
    Molecular Formula C8H17N2NaO6S2·xH2O
    Formula Weight 324.34g/mol (Anhydrous)
    MDL Number MFCD00065472
    Merck Index 14,7479
    Synonym Piperazine-1,4-bis(ethanesulfonic acid) monosodium salt
    TSCA TSCA
    Chemical Name or Material PIPES monosodium salt hydrate
    Assay 99%

    Description

    Applications
    PIPES [piperazine-N,N ft.-bis(2-ethanesulfonic acid)] is frequently used as a buffering agent in biochemistry. It is useful in cell culture work. It has been documented to minimize lipid loss when buffering glutaraldehyde histology in plant and animal tissues.

    Solubility
    Soluble in water

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit