PIPES, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Manufacturer: J.T.Baker 426500

Packaging POLYSTORMOR
Quantity 25g

 View more versions of this product

Catalog No. 02-003-625

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 5625-37-6
    Molecular Formula C8H18N2O6S2
    Formula Weight 302.37
    Synonym piperazine-N,N'-bis(2-ethanesulfonic acid)
    Chemical Name or Material PIPES
    Grade Ultrapure Bioreagent

    Specifications

    Formula Weight 302.37
    CAS 5625-37-6
    Synonym piperazine-N,N'-bis(2-ethanesulfonic acid)
    Quantity 25g
    Molecular Formula C8H18N2O6S2
    Packaging POLYSTORMOR
    Chemical Name or Material PIPES
    Grade Ultrapure Bioreagent

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit