Learn More
DWK Life Sciences KIMBLE™ Disposable Borosilicate Pasteur Pipette
SDP
KIMBLE™ Disposable Borosilicate Pasteur Pipette - These Pasteur-type pipettes are intended for one-time use in the transfer of small volumes.
Supplier:  DWK Life Sciences 8833500575
Catalog No. 12-141-547
Add to Cart
Description
Description
  • Designed to ASTM E732-80 Standard Specification for Disposable Pasteur-Type Pipette
  • Borosilicate pipets are manufactured from 51 expansion, low extractable borosilicate glass tubing conforming to USP Type 1 and ASTM E438, Type 1, Class B requirements
  • Soda lime pipets are manufactured from 90 expansion soda lime glass tubing conforming to USP Type III requirements
  • Ideal for the one-time use for small volume liquid transfer
  • Available either unplugged or with a cotton plug to suit most applications
  • Supplied both sterile and non-sterile
  • Blood bank pipettes (63C49 and 63C50) dispense 25 +/-3 drops per mL of serum or red cells at 23°C when held at 45°
  • Monstr-Pette™ (63DP1005) has a wider tip ID of 1.5mm and is ideally suited to more viscout samples
Specifications
Specifications
1.1 mm
Borosilicate Glass
2.25 in.
2 mL
7 mm
5.75 in.
250/Cs.
Product Suggestions
Product Suggestions