Learn More
Gilson™ PIPETMAN Classic™ Pipettes PROMO
The PIPETMAN Classic line, the world standard in pipetting, offers a wide range of continuously adjustable air-displacement pipettes suited for aqueous solutions of moderate viscosity and density.
Specifications
Material Polypropylene
Autoclavable Autoclavable
View More Specs
Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
F123600GPM
View Documents
Gilson™
F123600PM
2 to 20 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
F123601GPM
View Documents
Gilson™
F123601PM
20 to 200 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
F123602GPM
View Documents
Gilson™
F123602PM
200 to 1000 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Fully adjustable, air-displacement pipette with selected volume shown on digital indicator (volumeter)
  • Tip holder and tip ejector are autoclavable to avoid cross-contamination
  • Each pipette bears engraved identification number for traceability
Specifications
Specifications
Polypropylene
Autoclavable