Learn More
Gilson™ PIPETMAN™ Diamond Blister Pack Refill Pipette Tips
Autoclavable or gamma-ray sterlized
$100.50 - $369.50
Catalog Number | Mfr. No. | Volume (Metric) | Filtered | Sterility | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Filtered | Sterility | Price | Quantity & Availability | |||
F172511G
|
Gilson™
F172511 |
100 to 1000 μL | Non-Filtered | Sterile |
Pack of 960 for $100.50
|
|
F172510G
|
Gilson™
F172510 |
100 to 1000 μL | Non-Filtered | Non-sterile |
Pack of 960 for $100.50
|
|
F172313G
|
Gilson™
F172313 |
2 to 30 μL | Filtered | Sterile |
Pack of 960 for $369.50
|
|
F172311G
|
Gilson™
F172311 |
2 to 200 μL | Non-Filtered | Sterile |
Pack of 960 for $122.40
|
|
F172613G
|
Gilson™
F172613 |
100 to 1000 μL | Filtered | Sterile |
Pack of 960 for $369.50
|
|
F172513G
|
Gilson™
F172513 |
2 to 200 μL | Filtered | Sterile |
Pack of 960 for $176.00
|
|
F172310G
|
Gilson™
F172310 |
2 to 200 μL | Non-Filtered | Non-sterile |
Pack of 960 for $100.50
|
|
F172213G
|
Gilson™
F172213 |
0.1 to 10 μL | Filtered | Sterile |
Pack of 960 for $176.00
|
|
F172210G
|
Gilson™
F172210 |
0.1 to 20 μL | Non-Filtered | Non-sterile |
Pack of 960 for $100.50
|
|
Description
- Color-coded tip packaging corresponds to industry standards for volume color matching to pipettes
- Features graduations at varying volumes allowing for quick-checks when aspirating samples
- Standard or filtered tips
- Validated for use with 384-well tips: F172211G, F172311G and F17511G
Specifications
Clear | |
Pipetman™ Diamond™ | |
Polypropylene |