Learn More
Gilson™ PIPETMAN™ M Pipettes
Single or multichannel. Equipped with motorized piston that virtually eliminates pipetting variability among users and minimizes RSI (Repetitive Strain Injuries) among users
Manufacturer:  Gilson™ F81030
 View more versions of this product
Catalog No. F81030G
Add to cart
Description
Description
  • Motorized piston with adjustable piston speed that is compatible with varying solution densities
  • Lockable volume to prevent inaccuracies
  • Color-coded push-button corresponds to matching tip packs
  • Service alert reminders
  • Unique tip holder design allows for a secure fit of most brands of tips
  • Features four essential pipetting modes dash (en) reverse, mix, repetitive and pipetting to best fit your application
  • Pipette can be used while charging eliminating bottlenecks and downtime
  • Designed to exert equal pressure on each piston to ensure consistent volume levels across all channels
  • Power carrousel conveniently holds and charges up to 5 PIPETMAN M pipettes at once, charging 80% of its full capacity in less than one hour
  • Gilson Trident and unique spacer distribute ejection forces evenly over each tip, reducing the force required to eject tips
Specifications
Specifications
20 to 200µL
Yellow
12
D200, D300, DF100ST, DF200ST, DF300ST tips
1 Year
P12x200M
Pipette
Ergonomic