Learn More
DWK Life Sciences Wheaton™ Pipette Tips, Microtips, 350μL
High-quality non-sterile microtips made of natural-color polypropylene. Available in racks or bulk packs.
Manufacturer:  DWK Life Sciences W851190
 View more versions of this product
Catalog No. 02-912-681
  Check Availability
Add to cart
Description
Description
  • Made from high grade, virgin, metal-free polypropylene
  • Compatible with various pipette brands
  • Stringent quality procedures ensure lot to lot consistency
Specifications
Specifications
Universal Fit Pipettor
350μL
Natural
Universal Fit Pipettor
Rack
Non-sterile
PP (Polypropylene)