Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Pipette Tips, Microtips, 350μL
High-quality non-sterile microtips made of natural-color polypropylene. Available in racks or bulk packs.
Specifications
Volume (Metric) | 350 μL |
---|---|
Sterility | Non-sterile |
Color | Natural |
For Use With (Equipment) | Universal Fit |
Material | Polypropylene |
Catalog Number | Mfr. No. | Format | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Format | Quantity | Price | Quantity & Availability | ||||
02-912-680
|
DWK Life Sciences
W851189 |
Bulk | 1000 | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
02-912-681
|
DWK Life Sciences
W851190 |
Rack | 960 | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Made from high grade, virgin, metal-free polypropylene
- Compatible with various pipette brands
- Stringent quality procedures ensure lot to lot consistency
Specifications
350 μL | |
Natural | |
Polypropylene |
Non-sterile | |
Universal Fit |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title