Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pipsyl chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass Bottle
98-61-3
C6H4ClIO2S
302.51
MFCD00007441
POXFXTSTVWDWIR-UHFFFAOYSA-N
pipsyl chloride,4-iodobenzene-1-sulfonyl chloride,benzenesulfonyl chloride, 4-iodo,p-iodobenzenesulfonyl chloride,4-iodobenzenesulphonyl chloride,4-iodobenzenesulfonylchloride,4-iodophenylsulfonyl chloride,iodobenzene-p-sulfonyl chloride,unii-mvl274ezog,mvl274ezog
7399
4-iodobenzenesulfonyl chloride
C1=CC(=CC=C1S(=O)(=O)Cl)I
98-61-3
C6H4ClIO2S
302.51
MFCD00007441
POXFXTSTVWDWIR-UHFFFAOYSA-N
pipsyl chloride,4-iodobenzene-1-sulfonyl chloride,benzenesulfonyl chloride, 4-iodo,p-iodobenzenesulfonyl chloride,4-iodobenzenesulphonyl chloride,4-iodobenzenesulfonylchloride,4-iodophenylsulfonyl chloride,iodobenzene-p-sulfonyl chloride,unii-mvl274ezog,mvl274ezog
7399
4-iodobenzenesulfonyl chloride
C1=CC(=CC=C1S(=O)(=O)Cl)I
Packaging Glass Bottle
Melting Point 80°C to 84°C
Color Beige to White or Orange
Infrared Spectrum Authentic
Quantity 25g
Assay Percent Range 97%
Linear Formula IC6H4SO2Cl
Beilstein 11, 65
Merck Index 15, 7599
Formula Weight 302.51
Physical Form Crystals or Crystalline Powder
Percent Purity 97%
Chemical Name or Material Pipsyl chloride, 97%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-686-9

TSCA : TSCA