Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Pirenzepine dihydrochloride, 99%

Click to view available options
Quantity:
100mg
1g
5g
29868-97-1
C19H23Cl2N5O2
424.326
MFCD00055214
FFNMBRCFFADNAO-UHFFFAOYSA-N
pirenzepine dihydrochloride,pirenzepine hydrochloride,tabe,bisvanil,leblon,maghen,pirenzepine hcl,pirenzepine 2hcl,ls 519 dihydrochloride,unii-10ym403fls
71405
CHEBI:32014
11-[2-(4-methylpiperazin-1-yl)acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one;dihydrochloride
CN1CCN(CC1)CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4.Cl.Cl
29868-97-1
C19H23Cl2N5O2
424.326
MFCD00055214
FFNMBRCFFADNAO-UHFFFAOYSA-N
pirenzepine dihydrochloride,pirenzepine hydrochloride,tabe,bisvanil,leblon,maghen,pirenzepine hcl,pirenzepine 2hcl,ls 519 dihydrochloride,unii-10ym403fls
71405
CHEBI:32014
11-[2-(4-methylpiperazin-1-yl)acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one;dihydrochloride
CN1CCN(CC1)CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4.Cl.Cl

A M1 muscarinic receptor antagonist

Color White
Quantity 5g
Melting Point 192°C to 198°C
Sensitivity Hygroscopic
Merck Index 14,7491
Formula Weight 424.32
Physical Form Powder
Percent Purity 99%
Chemical Name or Material Pirenzepine dihydrochloride

EINECSNumber : 249-907-5

RTECSNumber : UU7883000

TSCA : No

Recommended Storage : Ambient temperatures